MaxQ™ Shaker Universal Platforms
MaxQ™ Shaker Universal Platforms
Actual product may vary
MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Dimensions (L x W)
30128Dual Stacked Universal Platform without Clamps, 18 x 18 in.MaxQ™ 2000, MaxQ™ 6000 Shaker18 x 18 in. (45.7 x 45.7 cm)
30115BIUniversal Platform without Clamps, 18 x 30 in.MaxQ 3000, 500030 x 18 in. (76 x 45.7 cm)
Catalog number 30128
Price (MYR)
7,028.00
EA
Add to cart
Description:
Dual Stacked Universal Platform without Clamps, 18 x 18 in.
For Use With (Equipment):
MaxQ™ 2000, MaxQ™ 6000 Shaker
Dimensions (L x W):
18 x 18 in. (45.7 x 45.7 cm)
Price (MYR)
7,028.00
EA
Add to cart
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Product TypeShaker Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)18 x 18 in. (45.7 x 45.7 cm)
Length (English) Platform18 in.
Length (Metric) Platform45.7 cm
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
DescriptionDual Stacked Universal Platform without Clamps, 18 x 18 in.
For Use With (Equipment)MaxQ™ 2000, MaxQ™ 6000 Shaker
Unit SizeEA