Search Thermo Fisher Scientific
| Catalog Number | Volume (Metric) Receiver |
|---|---|
| 300-4050 | 500 mL |
| 300-4000 | 250 mL |
| FIS09-740-23D | 500 mL |
| FIS09-740-23E | 1000 mL |
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing. 4 units per case.