15% Graphite/85% Vespel Ferrules
Product Image
15% Graphite/85% Vespel Ferrules
Mechanically robust for a long lifetime and are compatible with GC/MS
Have Questions?
Catalog NumberTemperature (Metric) Max
290VT155350°C
Catalog number 290VT155
Price (MYR)
-
Temperature (Metric) Max:
350°C
  • Form a strong grip with the column and cannot be reused as they form a permanant seal with the column
  • Temperature limit of 350°C, but must be retightened after initial temperature cycles
Specifications
DescriptionFerrules
For Use With (Application)GC/MS
Temperature (Metric) Max350°C
For Use With (Equipment)Thermo Scientific Trace Cold On-Column Injectors
Material15% Graphite, 85% Vespel
Unit SizeEach