SilberBrand™ ETERNA Type 2 Graduated Pipettes
SilberBrand™ ETERNA Type 2 Graduated Pipettes
Actual product may vary
SilberBrand™ ETERNA Type 2 Graduated Pipettes
SilberBrand™ ETERNA Type 2 Graduated Pipettes
SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Have Questions?
Catalog NumberVolume (Metric)ColorGraduations
270021 mLYellow0.01 mL
Catalog number 27002
Price (MYR)
-
Volume (Metric):
1 mL
Color:
Yellow
Graduations:
0.01 mL
  • DIN EN ISO 835
  • Calibrated to deliver (TD, Ex)
  • Specifications
    Diameter (Metric) Inner2.1, 3 mm
    For Use With (Equipment)Unifilter Direct-Connection HPLC Filter Systems
    CalibrationTo Deliver
    Certifications/ComplianceDIN EN ISO 835
    ClassB
    Diameter (Metric) Outer3.0/2.1 mm
    DescriptionUnifilter Direct-Connection Filter Holder; 2.1/3.0 mm
    Graduations0.01 mL
    Quantity12
    TypeUnifilter Direct Connection Holder
    ColorYellow
    Volume (Metric)1 mL
    Length (Metric)360 mm
    Unit SizeEach