Screw Top Vials with PTFE-lined Thermoset Cap
Screw Top Vials with PTFE-lined Thermoset Cap
Actual product may vary
Screw Top Vials with PTFE-lined Thermoset Cap
Suitable to inject samples from an autosampler, available in a variety of colors and volumes.
Have Questions?
Catalog NumberColorVolume (Metric)
27000Amber2 mL
Catalog number 27000
Price (MYR)
-
Color:
Amber
Volume (Metric):
2 mL
Specifications
Closure ColorGreen
DescriptionPre-assembled
Material (Liner)PTFE
Diameter (Metric) Inner4.6 mm
ColorAmber
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2 mL
No. per Pack100/Pk.
StyleStandard Opening
Closure MaterialMelamine Resin
Closure TypeSolid Top Cap
Height (Metric)32 mm
TypeScrew Top Vial Kit
Closure Size8-425
Unit SizeSet