TRACE™ GC Columns for Pesticides
TRACE™ GC Columns for Pesticides
Actual product may vary
TRACE™ GC Columns for Pesticides
Identify organophosphate and organochlorine pesticides with high performance, sensitivity and reproducibility.
Have Questions?
Catalog NumberStationary Phase
26RD142FTrace™ Pesticide II
Catalog number 26RD142F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Stationary Phase:
Trace™ Pesticide II
Identify organophosphate and organochlorine pesticides with high performance, sensitivity, and reproducibility using the Thermo Scientific™ TRACE™ GC Columns for Pesticides. These columns are specifically designed and tested to meet the most stringent requirements for the analysis of pesticides and herbicides.
Excellent Quality and Reproducibility:
  • Low bleed decreases MS contamination
  • Innovative design facilitates use at higher temperatures
  • Column inertness results in minimal peak tailing and decreased breakdown of sensitive samples
Alternative Uses:
  • Chlorinated aromatics
  • Plant sterols
  • Pyrethroids
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTRACE
Stationary PhaseTrace™ Pesticide II
USP TypeG43
Max. Temperature280/300°C
For Use With (Application)Pesticides Analysis
Particle Size0.2 μm
Film Thickness0.25 μm
Length (Metric)30 m
Description30 m x 0.25 mm x 0.25 um with 5 m Guard
Unit SizeEach