Determine methanol, fatty acid methyl esters (FAMEs) and glycerides in biodiesel analysis with high performance and reproducibility.
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric) Inner
Film Thickness
26AF024P
10 m
0.32 mm
0.1 μm
Catalog number 26AF024P
Price (MYR)
-
Length (Metric):
10 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.1 μm
Determine methanol, fatty acid methyl esters (FAMEs) and glycerides in biodiesel analysis with high performance and reproducibility using Thermo Scientific™ TRACE™ TR-BioDiesel GC Columns. Biodiesel is an important component in the development of carbon neutral fuels and specific GC columns within the TRACE range deliver robust performance to meet and exceed the methodology demands for this application area.
Columns Specifically Designed to Meet Your Biodiesel Analysis Needs:
TRACE TR-BioDiesel (M); for determination of residual methanol content in biodiesel according to EN Method 14110
TRACE TR-BioDiesel (G); for the analysis of total glyceride content according to EN Method 14105
TRACE TR-BioDiesel (F); for the analysis of FAME content in biodiesel according to EN Method 14103
TRACE TR-BioDiesel (G) ASTM; for the analysis of total glyceride content according to ASTM method D-6584