These Plain Packing Strips are made of odor-resistant, woven gauze fabric for use in wound care management applications such as nasal or sinus packing.
Have Questions?
Catalog Number
Dimensions (L x W)
262
also known as EM2IL62
5 yd. x 1 in.
Catalog number 262
also known as EM2IL62
Price (MYR)
2,050.00
CS
Add to cart
Dimensions (L x W):
5 yd. x 1 in.
Price (MYR)
2,050.00
CS
Add to cart
Made of a woven gauze fabric which reduces linting and fraying.
Iodoform provides odor-resistance to the packing strip.
Unbreakable poly bottles.
Tamper evident seals.
Used in wound care management applications such as nasal or sinus packing.