TRACE™ TR-5 GC Columns
TRACE™ TR-5 GC Columns
Actual product may vary
TRACE™ TR-5 GC Columns
Choose these excellent starting columns for method development, capable of performing most required separations.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
260E143P30 m0.32 mm0.25 μm
Catalog number 260E143P
Price (MYR)
-
Length (Metric):
30 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.25 μm
Excellent starting columns for method development,Thermo Scientific™ TRACE™ TR-5 GC Columns are capable of performing most required separations.
  • Nonpolar phase, 5% phenyl methyl polysiloxane
  • High operating temperature and extremely low-bleed
  • Widely used in a variety of applications
  • Similar to: DB-5, BP5, Rtx-5, HP-5, Ultra-2, PTE-5, SPB5, MDN-5, CP-Sil 8CB, SPB-5, AT-5, ZB-5, 007-2(MPS-5), SE-52, SE-54
Recommended for:
  • Alcohols
  • Free fatty acids
  • Aromatics
  • Flavors
  • Low polarity pesticides
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
Product LineTRACE
Stationary PhaseTrace™ TR-5
USP TypeG27, G36
Max. Temperature320/340°C
For Use With (Application)Free Fatty Acids Analyis, GC/MS for Free Fatty Acids, GC/MS for Aromatics, GC/MS for Flavors, GC/MS for Low Polarity Pesticides
Film Thickness0.25 μm
Length (Metric)30 m
PolarityNon-Polar
Description30 m x 0.32 mm x 0.25 μm
Unit SizeEach