TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard
TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard
Actual product may vary
TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard
The Thermo Scientific™ TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard offer the benefit of an integrated SafeGuard for extended column lifetime.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerParticle SizeIncludes
26096-142530 m0.25 mm0.25 μm5 m SafeGuard
Catalog number 26096-1425
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
30 m
Diameter (Metric) Inner:
0.25 mm
Particle Size:
0.25 μm
Includes:
5 m SafeGuard
Thermo Scientific™ TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard offer the benefit of an integrated SafeGuard for extended column lifetime.
Using a guard column with a TraceGOLD TG-5SILMS GC column will lead to greater consistency in results over a longer period of time.
  • With integrated SafeGUARD
  • Available in a range of phases
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-5 SILMS
USP TypeG27, G36
Max. Temperature330/350°C
For Use With (Application)GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons
Particle Size0.25 μm
Film Thickness0.25 μm
Length (Metric)30 m
PolarityLow-Polar
Includes5 m SafeGuard
Unit SizeEach