TraceGOLD TG-XLBMS GC Columns
TraceGOLD TG-XLBMS GC Columns
Actual product may vary
TraceGOLD TG-XLBMS GC Columns
Thermo Scientific™ TraceGOLD TG-XLBMS GC Columns offer a low polarity, low bleed column suitable for a wide range of applications.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
26079-154060 m0.25 mm0.25 μm
Catalog number 26079-1540
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
60 m
Diameter (Metric) Inner:
0.25 mm
Film Thickness:
0.25 μm
Choose these columns which offer a low-polarity, low-bleed column suitable for a wide range of applications.
  • Proprietary stationary phase
  • Unique selectivity
  • Similar to DB-XLB, Rxi-XLB, ZB-XLB, VF-XMS, MDN-12
Recommended for:
  • Pesticides
  • PCB congeners
  • PAH's
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-XLBMS
Max. Temperature340/360°C
For Use With (Application)Aroclor Mixes Analysis, GC/MS for PCB Congeners, GC/MS for PAHs, GC/MS for Aroclor Mixes
Film Thickness0.25 μm
Length (Metric)60 m
PolarityLow-Polar
Description60 m x 0.25 mm x 0.25 μm
Unit SizeEach