TraceGOLD™ TG-624SilMS GC Columns, 1.4μm film thickness, 0.25mm I.D., 30m length
TraceGOLD™ TG-624SilMS GC Columns, 1.4μm film thickness, 0.25mm I.D., 30m length
Actual product may vary
TraceGOLD™ TG-624SilMS GC Columns, 1.4μm film thickness, 0.25mm I.D., 30m length
Optimize volatile organics analysis with this low-bleed, midpolarity GC column with high thermal stability.
Have Questions?
Catalog NumberLength (Metric)
26059-332030 m
Catalog number 26059-3320
Price (MYR)
-
Length (Metric):
30 m
Optimizing volatile organics analysis requires a gas chromatography (GC) column with high thermal stability. Low-bleed, midpolarity Thermo Scientific™ TraceGOLD™ TG-624SilMS GC Columns offer the ideal performance and stability profile for this use.
Engineered for Superior Performance
  • Low to midpolarity phase, similat to 6% cyanopropylphenyl methylpolysiloxane
  • High thermal stability—maximum temperatures up to 320°C
  • Highly inert—excellent peak shape for a wide range of compounds
  • Equivalent to USP G43
Applications:
  • Residual solvents
  • Volatile organic compounds
  • Alcohols
  • Oxygenates
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • A low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended For:
  • Alcohols
  • Ketones
  • Residual solvents
  • Volatiles
Alternative Uses:
  • Oxygenates
Similar to these Columns:
  • DB-624
  • VF-624ms
  • CP-Select 624 CB
  • ZB-624
Specifications
Diameter (Metric)0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-624 SilMS
USP TypeG43
For Use With (Application)GC/MS
Film Thickness1.4 μm
Length (Metric)30 m
DescriptionTG-624 SilMS 30 m x 0.25 mm x 1.4 μm
QuantityPack of 1
Unit SizePack