Check Order Status
Air-Cushion Sole 2600 Shoes
Actual product may vary
Air-Cushion Sole 2600 Shoes
Antistatic with air cushion PU-sole and high slip resistance. Sanitized™ treated.
Have Questions?
Catalog Number
Size
260043
43
Catalog number
260043
Price (MYR)
2,215.00
KT
Add to cart
Size:
43
Price (MYR)
2,215.00
KT
Add to cart
CE EN ISO 20347:2012 O2 SRA
Grained leather
White
Slip resistance PU outsole sole
Replaceable insole (article no. 3551)
Sanitized treated
Antistatic
Lining microfiber, respiration absorbing
Instep area with lacing
Specifications
Certifications/Compliance
CE EN ISO 20347:2012 O2 SRA
Color
White
Liner Material
Microfiber
Material
Leather
Size
43
Sole Features
Slip-resistant
Sole Material
Polyurethane
Style
2600
Unit Size
KT