Hypersil™ GOLD HPLC Columns, 3μm Particle Size
Hypersil™ GOLD HPLC Columns, 3μm Particle Size
Actual product may vary
Hypersil™ GOLD HPLC Columns, 3μm Particle Size
Endcapped, ultrapure, silica-based columns with exceptional peak shape and resolution for HPLC and LC/MS
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
25003-254030250 mm4 mm
Catalog number 25003-254030
Price (MYR)
-
Length (Metric):
250 mm
Diameter (Metric):
4 mm
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data
Specifications
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)250 mm
Particle Size3 μm
pH1 to 11
Pore Size175 Å
Surface Area220 m²/g
USP TypeL1
Stationary PhaseC18
Packing MaterialSilica
Carbon Load10%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach