Nunc™ Petri Dishes
Nunc™ Petri Dishes
Actual product may vary
Nunc™ Petri Dishes
Nunc™ Petri Dishes
Nunc™ Petri Dishes
Nunc™ Petri Dishes
Prepare samples in hospital environments, food science or the pharmaceutical industry with these petri dishes
Have Questions?
Catalog NumberDiameter (Metric)
249964140 mm
Catalog number 249964
Price (MYR)
1,189.00
CS
Add to cart
Diameter (Metric):
140 mm
Price (MYR)
1,189.00
CS
Add to cart
Prepare samples in hospital environments, food science or the pharmaceutical industry with Thermo Scientific™ Nunc™ Petri Dishes. These dishes are available with a molded grid and perform well in automatic dispensers due to complete flatness and uniform height.
  • For the culturing of bacteria, fungi and other microorganisms
  • Constructed of high-quality and optically clear polystyrene, with lid
  • Fitted with top and bottom stacking rings for easier handing and stable stacking
  • Vented for gaseous exchange
  • Sterilized by gamma irradiation
Specifications
Bottom ShapeFlat
Culture Area145 sq. cm
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentNon-treated
TypePetri Dish
Well Count1
No. per Case80
No. per Pack10
Height (Metric)20 mm
Height (English)0.75 in.
Diameter (Metric)140 mm
Unit SizeCS