Check Order Status
Filter Discs with PTFE Membrane, 0.45 μm
Actual product may vary
Filter Discs with PTFE Membrane, 0.45 μm
Pack of filter discs with PTFE membranes.
Have Questions?
Catalog Number
Diameter (Metric)
24745-NP
47 mm
Catalog number
24745-NP
Price (MYR)
-
Diameter (Metric):
47 mm
Pack of filter discs with PTFE membranes.
Pack of filter discs with PTFE membranes.
47 mm diameter
Made of hydrophilic polypropylene
Pore Size: 0.45 μm
Specifications
Material
Polypropylene (Hydrophilic)
Filtration Area
0.45 μm
No. per Pack
100
Wettability
Hydrophilic
Diameter (English)
1.9 in.
Diameter (Metric)
47 mm