4301 Stainless Steel Spoon
4301 Stainless Steel Spoon
Actual product may vary
4301 Stainless Steel Spoon
Have Questions?
Catalog NumberLength (Metric)
242
also known as CP242
150 mm
Catalog number 242
also known as CP242
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
Specifications
Product TypeSpoon
For Use With (Application)For Chemical
Dimensions (L x W)22 x 30 mm
MaterialStainless Steel (4301)
Length (Metric)150 mm
Unit SizeCS