Check Order Status
Acrodisc™ Syringe Filters with wwPTFE Membrane and Minispike Outlet
Actual product may vary
Acrodisc™ Syringe Filters with wwPTFE Membrane and Minispike Outlet
Universal membrane syringe filters for all of your analytical filtration requirements.
Have Questions?
Catalog Number
Pore Size
Quantity
2401
also known as CAN BUY FROM MANU. CALL T.ARL FOR PT#
0.2 μm
1000/Pk.
Catalog number
2401
also known as CAN BUY FROM MANU. CALL T.ARL FOR PT#
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.2 μm
Quantity:
1000/Pk.
Universal membrane syringe filters for all of your analytical filtration requirements.
This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Material (Membrane)
wwPTFE
Pore Size
0.2 μm
Sterility
Non-sterile
Filtration Area
1 cm
2
Hold up Volume (After Air Purge)
<14 μL
Housing Material
Polypropylene
Outlet Style
Minispike Outlet
Quantity
1000/Pk.
Temperature (Metric) Max
55°C
Diameter (Metric)
13 mm