Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Actual product may vary
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
236099Flask Clamp Starter Kit for MaxQ High Performance ShakersMaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Catalog number 236099
Price (MYR)
5,296.00
KT
Add to cart
Description:
Flask Clamp Starter Kit for MaxQ High Performance Shakers
For Use With (Equipment):
MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Price (MYR)
5,296.00
KT
Add to cart
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
Specifications
Product TypeFlask Clamp Kit
DescriptionFlask Clamp Starter Kit for MaxQ High Performance Shakers
For Use With (Equipment)MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Unit SizeKT