HyPURITY™ AQUASTAR™ Guard Cartridges
HyPURITY™ AQUASTAR™ Guard Cartridges
Actual product may vary
HyPURITY™ AQUASTAR™ Guard Cartridges
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerParticle Size
22503-0130013 mm3 μm
HYP22503-0130013 mm3 μm
Catalog number 22503-013001
Price (MYR)
-
Diameter (Metric) Inner:
3 mm
Particle Size:
3 μm
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime.
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holder
  • Polar endcapped offer alternative selectivity
  • High quality, highly pure silica columnss
Specifications
Particle Size3 μm
Length (Metric)10 mm
Diameter (Metric) Inner3 mm
Stationary PhaseC18
Product LineHyPURITY
Unit SizeEach