Flat Bottom Polystyrene Multipurpose Container with Metal Screw Cap
Flat Bottom Polystyrene Multipurpose Container with Metal Screw Cap
Actual product may vary
Flat Bottom Polystyrene Multipurpose Container with Metal Screw Cap
Ideal for optical measurements due to its high clarity.
Have Questions?
Catalog NumberCapacity (Metric)SterilityLabel
224180100 mLAsepticLabeled
Catalog number 224180
Price (MYR)
2,660.00
KT
Add to cart
Capacity (Metric):
100 mL
Sterility:
Aseptic
Label:
Labeled
Price (MYR)
2,660.00
KT
Add to cart
  • Metal screw cap
  • With or without plain label
  • Manufactured aseptically in a Class II Clean Room environment
  • Can be used universally for academic and medical purposes
Specifications
Product TypeMulti-purpose Container
MaterialPolystyrene
SterilityAseptic
Capacity (Metric)100 mL
Closure MaterialMetal
Closure TypeScrew Cap
Diameter (Metric)49 mm
For Use With (Application)For academic and medical purposes
LabelLabeled
Length (Metric)80 mm
Unit SizeKT