Evolution™ Specular Reflectance Accessories
Evolution™ Specular Reflectance Accessories
Actual product may vary
Evolution™ Specular Reflectance Accessories
Get high performance and accuracy for the measurement of glossy or mirrored surfaces at fixed angles with Thermo Scientific™ Evolution™ Specular Reflectance Accessories.
Have Questions?
Catalog NumberIncludes
222-23960015° Specular Reflectance
Catalog number 222-239600
Price (MYR)
78,577.00
EA
Add to cart
Includes:
15° Specular Reflectance
Price (MYR)
78,577.00
EA
Add to cart
Measure at 8, 15, 20, 30, 45 and 60 degrees, or at any angle from 30 degrees to 80 degrees, with Thermo Scientific™ Evolution™ Specular Reflectance Accessories.
This suite of accessories is useful for measuring film thickness and reflectance properties from coatings on metallic, glass and silicon substrates. The accessory includes an alignment mirror that can also be used for baseline measurements.
Convenient Horizontal Sample Mounting
  • Convenient horizontal sample mounting allows easy positioning of samples for measurement
  • Selection of apertures limits size of the light patch striking the sample surface, facilitating reproducible sampling
  • Align it once and it's aligned for life- no need to align each time you insert the accessory
Use 15-60 degree accessories together with a polarizer or filter
  • The small, fixed angle accessories can be used together with a polarizer, depolarizer or filter holder to modify the light beam before or after striking the test mirror
  • The Evolution Solid Sample Slide Holder accommodates multiple slide holders, allowing optical elements to be inserted either before or after the Fixed-Angle Specular Reflectance accessory
Use for QA/QC Testing
  • Designed for applications where testing at specific angles is required
  • Test reflectance of each piece quickly and efficiently to ensure that production specification requirements are met
*UV-Vis VeeMax accessory is trademarked by Pike Technologies
Specifications
For Use With (Equipment)Thermo Scientific Evolution Pro Spectrophotometer
Includes15° Specular Reflectance
Unit SizeEA