FID Maintenance Kits
FID Maintenance Kits
Actual product may vary
FID Maintenance Kits
For use with Agilent GCs
Have Questions?
Catalog NumberFor Use With (Equipment)
22180Agilent 5890 GCs
Catalog number 22180
Price (MYR)
-
For Use With (Equipment):
Agilent 5890 GCs
  • Includes a variety of the most common consumable GC supplies and tools
  • All parts meet or exceed performance of instrument manufacturer's parts
  • Parts list makes reordering easy
Specifications
TypeFID Maintenance Kit
Particle Size0.2 μm
Type2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
Diameter (Metric) Inner2.1 mm
IncludesGraphite Ferrules (1/4 in., 0.4/0.5/0.8 mm ID), FID/NPD Capillary Adaptor, Capillary Nuts, Jet Reamers/Ferrule Removers, Nut (1/4 in.), Scoring Wafer, Capillary Column Caps, Ignitor, FID Flow-Measuring Adaptor, Wrench (1/4 x 5/16 in.), Installation Gauge, Wire Cleaning Brush, High-Performance Siltek™ Treated FID Adaptable Jet, Spanner Wrench, FID Jet Removal Tool
Product LineThermo Scientific
Quantity1 Kit
For Use With (Equipment)Agilent 5890 GCs
Unit SizeEach