Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Actual product may vary
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Freeze samples effectively with impact-resistant these narrow-mouth, polycarbonate bottles with closure.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
2205-00321 L38-430 mm
Catalog number 2205-0032
Price (MYR)
1,827.00
CS
Add to cart
Capacity (Metric):
1 L
Closure Size:
38-430 mm
Price (MYR)
1,827.00
CS
Add to cart
Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure, which are window clear. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features:
  • 16 oz. and 0.25 gal. sizes are good for smaller volume applications where contents need to be seen
  • 0.5 and 0.66 gal. sizes can be used as culture vessels for batch, suspension or nonattached roller culturing
  • 1 and 2 gal. sizes have built-in shoulder loops for attaching an identification tag
  • Excellent impact resistance makes these bottles good for freezer use to -135°C (-211°F)
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Transparent/Leakproof† when used with Nalgene closures
Includes:
Leakproof† polypropylene screw caps
Note:
Before autoclaving, completely disengage threads or remove cap. Autoclave empty. Repeated autoclaving will shorten the bottle life.
.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity32 oz., 1 L
Capacity (Metric)1 L
Capacity (English)32 oz.
ColorClear
DescriptionNarrow-Mouth Bottle
Diameter (Metric) Inner Neck27 mm
MaterialPolycarbonate
No. per Case24
No. per Pack4
SterilityNon-sterile
Closure ColorNatural
Neck Finish38 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1580 for food and beverage use, USP Class VI.
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)221 mm
Product TypeNarrow Mouth Bottle
Width (English)3.64 in.
Height (English)9.05 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Diameter (Metric)99 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS
Frequently asked questions (FAQs)
Why are your Nalgene containers and caps always different materials?
In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.