ART™ Barrier Specialty Pipette Tips, 200 XLG
ART™ Barrier Specialty Pipette Tips, 200 XLG
Actual product may vary
ART™ Barrier Specialty Pipette Tips, 200 XLG
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Catalog NumberVolume (Metric)
2160G200 μL
Catalog number 2160G
Price (MYR)
4,305.00
PK
Add to cart
Volume (Metric):
200 μL
Price (MYR)
4,305.00
PK
Add to cart
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available racked, with 96 tips per tray (8 trays per pack)
  • Manufactured according to the industry's highest quality standard
  • Designed to aid in pipetting for a variety of applications
  • Extended length prevent contamination by keeping the pipette shaft from entering the sample vessel
  • Universal fit
Specifications
DescriptionART XLG, Filtered, Sterile,
Volume (Metric)200 μL
FilteredFiltered
SterilitySterile
ModelART XLG
Length (English)3.59 in.
Length (Metric)9.12cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleExtended Length, Genomic/Wide Orifice
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePK