Eliminate sample retention with these specially designed pipette tips.
Have Questions?
Catalog Number
Volume (Metric)
Model
Tip Style
Fit for Purpose
2149P-05
20 μL
ART 20P
Low Retention, Micro Point
Sterile and PCR Ready
Catalog number 2149P-05
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
20 μL
Model:
ART 20P
Tip Style:
Low Retention, Micro Point
Fit for Purpose:
Sterile and PCR Ready
Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.
Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
Crystal clear tips allow you to easily see transparent samples
Tips seal when exposed to potential contaminants
Certified RNase, DNase, ATP, and Endotoxin Free
Specifications
DescriptionART 20P, Filtered, Sterile, Rack
Volume (Metric)20 μL
FilteredFiltered
SterilitySterile
ColorClear
AutoclavableNot Autoclavable
ModelART 20P
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free