SafeBasics™ Procedure Earloop Face Masks
SafeBasics™ Procedure Earloop Face Masks
Actual product may vary
SafeBasics™ Procedure Earloop Face Masks
Ultrasonically sealed procedure face masks are soft and stretchy with latex-free earloops and an adjustable nosepiece to ensure a comfortable fit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorASTM Level
2140WhiteASTM Level 2
2130
also known as STARA2130
WhiteASTM Level 1
Catalog number 2140
Price (MYR)
4,560.00
PK
Add to cart
Color:
White
ASTM Level:
ASTM Level 2
Price (MYR)
4,560.00
PK
Add to cart
  • Primary protection for simple, low-risk procedures involving very light splatter, moisture, spray and spatter. Ideal for patient appointments and exams
  • Inner and outer layer constructed for basic fluid resistance, with ultrasonically sealed, soft, stretchy, latex free earloops
  • Adjustable nosepiece for custom and comfortable fit and easy breathing
  • Economic alternative for simple procedures
  • ASTM Level 1, 2 and 3 protection
Specifications
Product TypeMedical/Procedural Face Mask
ASTM LevelASTM Level 2
ColorWhite
Fastening TypeEarloops
MaterialSpunbond Polypropylene
SizeStandard
SterilityNon-sterile
StylePleated
Certifications/ComplianceASTM F2299, ASTM F1862
Certifications/Compliance (Europe)EN 14683
Construction3-ply
DisposableSingle-use
Filter Efficiency≥98% (PFE), ≥98% (BFE)
For Use With (Application)Animal health, health and wellness, medical, industrial, laboratory and safety
Latex FreeYes
Model2140
Packaging Quantity50 Masks/Box, 10 Boxes/Cs.
Pore Size0.1 μm
SeriesSafeBasics™
Unit SizePK