Isotemp™ R20 Refrigerated and Heated Bath Circulators: 5.4-6.5L, 230V/50Hz
Isotemp™ R20 Refrigerated and Heated Bath Circulators: 5.4-6.5L, 230V/50Hz
Actual product may vary
Isotemp™ R20 Refrigerated and Heated Bath Circulators: 5.4-6.5L, 230V/50Hz
For laboratory and industrial applications.
Have Questions?
Catalog NumberModelType
211131100Isotemp 4100 R20Tall Form
Catalog number 211131100
Price (MYR)
24,528.00
EA
Add to cart
Model:
Isotemp 4100 R20
Type:
Tall Form
Price (MYR)
24,528.00
EA
Add to cart
Low-profile model enables easy bath access when lab counter height presents a challenge. Seamless stainless-steel tanks guarantee durability and easy cleaning. Robust refrigeration system ensures that samples and applications can be cooled quickly. Fine-tuned refrigeration metering allows for excellent temperature stability at all times.
  • All bath drains are located on the front of the unit with an integrated valve
Green Tip: For less demanding applications, power consumption can be lowered by utilizing the energy savings mode
Specifications
Cooling Capacity250 W
Pump TypeForce
Temperature Range-20°C to +100°C
TypeTall Form
Heating Capacity1.2 kW
Max. Pressure17 L/min.; 310 mbar
ModelIsotemp 4100 R20
Temperature Stability±0.05°C
Weight (English)58.0 lb.
Weight (Metric)26.3 kg
Voltage230 V
Frequency50 Hz
Unit SizeEA