Mini-Prep Isolation of Plasmid DNA
Mini-Prep Isolation of Plasmid DNA
Actual product may vary
Mini-Prep Isolation of Plasmid DNA
Introduces the principles of extracting plasmid DNA from bacterial cells.
Have Questions?
Catalog number 202
Price (MYR)
-
Small-scale rapid isolation of plasmid DNA is a routine procedure used for screening and analysis of recombinant DNA in cloning and subcloning experiments.
  • In this experiment, students isolate plasmid DNA without the use of toxic chemicals like phenol or chloroform
  • Complete in 1 hour 45 minutes
Specifications
Bulb Draw (Metric)3.5 mL
DescriptionGeneral Purpose Transfer Pipettes, Standard Bulb, 7.7mL
Diameter (Metric) Outer7.9 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL21
GraduatedNo
FormatKit
Isolation TechnologyOrganic Extraction
Packaging TypeBulk 500/Pk
Volume (Metric)7.7 mL
Length (Metric)15.6 cm
TypeGeneral Purpose Transfer Pipet
DisposableSingle-use
For Use With (Equipment)Agarose Gel electrophoresis
For Use With (Application)Use for screening and analysis of recombinant DNA in cloning and subcloning experiments
IncludesInstructions, plasmid extraction LyphoCells™, Gel-loading solution, UltraSpec-Agarose™ Powder, electrophoresis Buffer, InstaStain™ FlashBlue™ Stain, FlashBlue Plus™
Product TypeIsolation of Plasmid DNA Kit
Quantity1/Pk.
Unit SizeEA