Accessories for MaxQ™ 8000 Shakers
Accessories for MaxQ™ 8000 Shakers
Actual product may vary
Accessories for MaxQ™ 8000 Shakers
Accessories for MaxQ™ 8000 Shakers
Accessories for MaxQ™ 8000 Shakers
Accommodate various applications and budgets with these flexible and convenient Thermo Scientific™ MaxQ™ 8000 Shaker Accessories.
Have Questions?
Catalog NumberDescription
194223Stand for two stackable shakers
Catalog number 194223
Price (MYR)
12,491.00
EA
Add to cart
Description:
Stand for two stackable shakers
Price (MYR)
12,491.00
EA
Add to cart
Accommodate various applications and budgets with these flexible and convenient Thermo Scientific™ MaxQ™ 8000 Shaker Accessories. With exceptional simplicity and flexibility, these shaker accessories are the optimal choice for busy labs.
Specifications
Product TypeStand
Depth (English)30.5 in.
Depth (Metric)75.2 cm
Height (English)11.8 in.
Height (Metric)30 cm
Width (English)45.5 in.
Width (Metric)115.6 cm
Weight (English)142 lb.
DescriptionStand for two stackable shakers
For Use With (Equipment)MaxQ 8000
Unit SizeEA