Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Actual product may vary
Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
Choice of accessories for maximum flexibility
Have Questions?
Change view
Catalog Number
Product Type
1910179
Hanging Shelf
1910112
Replacement SmartPort Grommets
2 Options
Catalog number 1910179
Price (MYR)
-
Product Type:
Hanging Shelf
Choice of accessories for maximum flexibility
UV Light
Provides safe and easy method for cabinet disinfection
Factory-installed; order with cabinet
Included with packages
Armrest and Footrest
Stainless-steel armrests (set of two) provide ergonomic forearm support and extended reach inside cabinet with less airflow disruption; included with packages
Footrest provides ergonomic foot and posture support during seated use
IV Bag Holder Kit
Bar and 12 hooks
For hanging IV bags near interior ceiling of cabinet
Valves
For standard applications or universal piping
Standard-application valves have long stem pipe fitting appropriate for side-access media services
Short piping stem on universal-piping valves will not penetrate cabinet side wall
Combustible gas valve: Rated for combustible gas dispensing inside cabinet chamber
Noncombustible gas valve: Rated for noncombustible gas dispensing inside cabinet chamber
Vacuum valve: Rated for routing vacuum inside cabinet chamber
Thimble Exhaust Duct Transition
Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
Method recommended by NSF/ANSI™ 49 for externally exhausting Class II, Type A2 biological safety cabinets
Universal Piping for Exhaust Systems
Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
Factory-installed; must be ordered with cabinet
Specifications
Certifications/ComplianceNSF/ANSI 49, Class II Type A2