Cryo Aprons
Cryo Aprons
Actual product may vary
Cryo Aprons
Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189453Large
189452Medium
Catalog number 189453
Price (MYR)
2,593.00
EA
Add to cart
Size:
Large
Price (MYR)
2,593.00
EA
Add to cart
Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.
  • Small: 36 in. (91.4cm)
  • Medium: 42 in. (106.7cm)
  • Large: 48 in. (121.9cm)
  • Warranty varies by country, please contact us for details
Specifications
TypeCryogenic Apron
SizeLarge
ColorBlue
Width (English)48 in.
Width (Metric)121.9 cm
Unit SizeEA