Waterproof Cryo Gloves - Elbow Length
Waterproof Cryo Gloves - Elbow Length
Actual product may vary
Waterproof Cryo Gloves - Elbow Length
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189447X-Large
189445Medium
189446Large
Catalog number 189447
Price (MYR)
3,636.00
PAA
Add to cart
Size:
X-Large
Price (MYR)
3,636.00
PAA
Add to cart
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
  • Side release buckles for quick on/off
Specifications
DescriptionElbow Length
Product TypeWaterproof Cryogenic Gloves
SizeX-Large
For Use With (Application)Cryogenic Preservation
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizePAA