CD105 Rat anti-Mouse, Unconjugated, Clone: MJ7/18, Southern Biotech™
Product Image
CD105 Rat anti-Mouse, Unconjugated, Clone: MJ7/18, Southern Biotech™
Rat Monoclonal Antibody
Have Questions?
Catalog number 186001
Price (MYR)
-
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
AntigenCD105
ApplicationsFlow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
ClassificationMonoclonal
CloneMJ7/18
Concentration0.5mg/mL
ConjugateUnconjugated
FormulationSupplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
Host SpeciesRat
ImmunogenInflamed mouse skin
Quantity0.5 mg
Regulatory StatusRUO
Primary or SecondaryPrimary
Target SpeciesMouse
FormLiquid
IsotypeIgG2a
Unit SizeEA
Contents & Storage
Store at 2-8°C