Accucore™ 150 Amide HILIC HPLC Columns
Accucore™ 150 Amide HILIC HPLC Columns
Actual product may vary
Accucore™ 150 Amide HILIC HPLC Columns
Designed for the separation of hydrophilic biomolecules in HILIC mode—an excellent choice for glycan separations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
16726-154630
also known as 16726-154630B
150 mm4.6 mm2.6 μm
16726-152130
also known as 16726-152130B
150 mm2.1 mm2.6 μm
16726-01210510 mm2.1 mm2.6 μm
Catalog number 16726-154630
also known as 16726-154630B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Particle Size:
2.6 μm
Designed for the separation of hydrophilic biomolecules in HILIC mode, Thermo Scientific™ Accucore™ 150 Amide HILIC LC Columns are an excellent choice for glycan separations.Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations—without the elevated backpressures required by sub-2μm particles.
Rugged and reproducible 2.6μm solid core particles.
  • Fast separations with superb resolutionLow backpressures
  • Amide phase bonded onto 150Å pore diameter solid core particles
  • High retention of a broad range of hydrophilic analytes in HILIC mode
  • Recommended for hydrophilic biomolecules such as glycans
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid-core
Particle Size2.6 μm
pH2 to 9
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeN/A
Stationary PhaseAmide
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeHILIC
PhaseHILIC
TechniqueBio-LC
Unit SizeEach