Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Actual product may vary
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Achieve excellent cell attachment and growth rates with Nunc MiniTrays with Nunclon Delta Surface.
Have Questions?
Catalog NumberWell Count
16311860
Catalog number 163118
Price (MYR)
3,116.00
CS
Add to cart
Well Count:
60
Price (MYR)
3,116.00
CS
Add to cart
Achieve excellent cell attachment and growth rates with Thermo Scientific™ Nunc™ MiniTrays with Nunclon™ Delta Surface.
  • Sterile polystyrene
  • Available with 60 or 72 conical wells
  • Large raised alphanumeric coordinates
  • Keyed lid for proper orientation
Specifications
Certifications/ComplianceCertified Nunclon surface
Closure IncludedYes
Bottom ShapeConical
Capacity (Metric)8 L
Culture Area0.013 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentNunclon Delta
TypeMini Tray
Volume (Metric) Working8 μL
Well Count60
IncludesLid
No. per Case150
No. per Pack10
Length (Metric)84.1 mm
Width (Metric)5.82 mm
Height (Metric)11.2 mm
Unit SizeCS
Frequently asked questions (FAQs)
What reagents are used to coat your Nunclon Delta surface products?
The Nunclon Delta surface is a surface modification that enhances cell attachment and growth for adherent cell lines and is not coated with any chemical reagent.