Racks and Inserts for Refrigerated and Heated Bath Circulators
Racks and Inserts for Refrigerated and Heated Bath Circulators
Actual product may vary
Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)Holds
160-0079Stainless-steel rackBath Types A25B, A40, S21, S30Depends on insert (purchase separately)
Catalog number 160-0079
Price (MYR)
1,615.00
EA
Add to cart
Description:
Stainless-steel rack
For Use With (Equipment):
Bath Types A25B, A40, S21, S30
Holds:
Depends on insert (purchase separately)
Price (MYR)
1,615.00
EA
Add to cart
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
DescriptionStainless-steel rack
For Use With (Equipment)Bath Types A25B, A40, S21, S30
TypeRack
HoldsDepends on insert (purchase separately)