AC150 Immersion Circulators
AC150 Immersion Circulators
Actual product may vary
AC150 Immersion Circulators
AC150 Immersion Circulators
AC150 Immersion Circulators
AC150 is ideal when applications require greater pumping performance, ramp programming, application alarms and temperature ranges from -50° to +150°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelVoltageFrequency
155-0021AC150 with bridge230 V50 Hz
155-0001AC150230 V50 Hz
Catalog number 155-0021
Price (MYR)
14,024.00
EA
Add to cart
Model:
AC150 with bridge
Voltage:
230 V
Frequency:
50 Hz
Price (MYR)
14,024.00
EA
Add to cart
Thermo Scientific™ AC150 is ideal when applications require greater pumping performance, ramp programming, application alarms and temperature ranges from 13° to 200°C. Features include an enhanced digital display and easy–to–use touchpad settings, allowing you to easily tune the temperature to meet your specific application requirements.
  • Max. temperature: 200°C
  • Low-liquid level alarm for Safety Class III
  • Automatic controller shutdown at detection of excessive high temperature, low liquid level or motor overload
  • USB communication port and options for RS-232, RS-485, Ethernet/LAN, Analog I/O
  • Programmable application temperature alarm with user selected alarm, go-safe-state or shut off option
  • Self-Tuning PID controller for optimized temperature control
  • Five programmable set point temperatures
  • RTA (Real Temperature Adjustment) for calibration
  • Internal or external temperature control mode (Remote Sensor, NAMUR type)
  • Pump speed adjustment to three levels for turbulance control
  • Powerful force and suction pump for external open and closed applications
  • Three languages (English, German, French)
  • Change digital display resolution between 0.1 and 0.01 and between °C — °F — °K
  • Acoustic and optical alarm
  • Auto-Restart feature after power failure
Includes:8mm and 12mm hose barbs with bridge (–0026, –0029, –0021 only), pump plug for external circulation, 6ft. power cord.
  • Warranty varies by country, please contact us for details
Specifications
DescriptionWith bridge; 230V/50Hz
Certifications/ComplianceCE marked, UL listed, CSA, RoHS
CommunicationsUSB, RS232, RS485, Ethernet/LAN, Analog I/O
Flow Rate20 L/min.
Temperature Range13°C to 200°C
Heating Capacity2 kW/1.2 kW
Max. Pressure475 mbar/6.89 psi
Max. Suction330 mbar/4.85 psi
ModelAC150 with bridge
Safety Class3/FL
Temperature Stability0.01°C
Dimensions (L x W x H)7.8 x 6.4 x 14.6 in. (19.9 x 16.5 x 37.2 cm)
Length (English)7.8 in.
Length (Metric)19.9 cm
Width (English)6.4 in.
Width (Metric)16.5 cm
Height (English)14.6 in.
Height (Metric)37.2 cm
Weight (English)9.25 lb.
Weight (Metric)4.2 kg
Electrical Requirements230 V 50 Hz
Voltage230 V
Frequency50 Hz
Unit SizeEA