SAHARA PPO S5P Heated Bath Circulators
SAHARA PPO S5P Heated Bath Circulators
Actual product may vary
SAHARA PPO S5P Heated Bath Circulators
SAHARA PPO S5P Heated Bath Circulators
SAHARA PPO S5P Heated Bath Circulators
Ideal when visibility of your application is required.
Have Questions?
Catalog NumberController TypeVoltageFrequency
152-3051SC100230 V50 Hz
Catalog number 152-3051
Price (MYR)
10,972.00
EA
Add to cart
Controller Type:
SC100
Voltage:
230 V
Frequency:
50 Hz
Price (MYR)
10,972.00
EA
Add to cart
Ideal when visibility of your application is required. Thermo Scientific SAHARA PPO S5P Heated Bath Circulators maintain temperatures from ambient +13° to 100°C, with maximum bath volume of 5 liters. The SAHARA series of heated bath circulators are available with a wide variety of work area dimensions to meet your internal or external circulation requirements. A selection of eight different controllers will create the perfect fit for your application. Rugged and corrosion-resistant for high temperature applications up to 200°C.
Environment-friendly Design: Save up to 80% on energy costs and thousands of kilowatt hours during the life of the system. All units are RoHS-compliant (lead- and mercury-free) and use recycled packaging.
Safe Operation: All units are UL/CSA compliant, assuring safe operation in the unlikely event of a fault condition. 115/60 Hz units are UL listed or recognized, and all units are UL/CSA and RoHS compliant. IQ/OQ compliance is optional.
Ease of Use: All thermostats feature an intuitive user interface with bright display to view critical readings. Each system comes with a quick-start guide for simple setup and operation. Thermostat can be turned and positioned on all four sides of the bath for easy viewing.
Superior Warranty and Service: All solutions come with a 36-month warranty and are backed by Thermo Scientific service and support worldwide.
  • Capacity: 5L
  • Modified polyphenylene oxide baths are thermally resistant up to 100°C
  • Deliver exceptional temperature performance with operational savings
  • Temperatures are maintained from ambient plus 13° to 100°C
SC 100 controller
  • Features 3 languages, acoustic and optical alarms, and five user-settable programs
  • Two pump speeds allow adjustement of water flow within bath to reduce turbulence
SC 150 controller
  • Additional features include a USB port, a low-liquid level alarm, and an automatic controller shut-down at detection of excessive high temperature, low liquid level, or motor overload
Includes:
Communication cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation, 6ft. power cord
Recommended for:
  • Calibration
  • Bioreactors
  • Rotary Evaporators
  • Condensers
  • Sample/Material Testing
  • Sample/Material Preparation
  • Warranty varies by country, please contact us for details
Specifications
Controller TypeSC100
Capacity (Metric)5 L
Certifications/ComplianceCE/ROHS/WEEE
CommunicationsUSB
Flow Rate17 L/min.
Temperature RangeAmbient +13° to 300°C
Dimensions (L x W x H)15.3 x 7.5 x 14.2 in. (38.8 x 19 x 36 cm)
Length (English)15.3 in.
Length (Metric)38.8 cm
Width (English)7.5 in.
Width (Metric)19 cm
Height (English)14.2 in.
Height (Metric)36 cm
Weight (English)11.2 lb.
Weight (Metric)5.1 kg
Electrical Requirements230 V 50 Hz
Voltage230 V
Frequency50 Hz
Unit SizeEA