1300 Series Class II, Type A2 Biological Safety Cabinet Packages, 230V 50/60Hz
1300 Series Class II, Type A2 Biological Safety Cabinet Packages, 230V 50/60Hz
Actual product may vary
1300 Series Class II, Type A2 Biological Safety Cabinet Packages, 230V 50/60Hz
Perform daily applications safely and efficiently with this simple-to-order package, which includes a cabinet, stand, pre-installed UV light and armrests.
Have Questions?
Catalog NumberChamber MaterialWidth (Metric) ExteriorSash Opening (Metric)Description
1396SmartCoat130 cm20.3 cm20.3 cm Window
Catalog number 1396
Price (MYR)
-
Chamber Material:
SmartCoat
Width (Metric) Exterior:
130 cm
Sash Opening (Metric):
20.3 cm
Description:
20.3 cm Window
Perform daily applications safely and efficiently with simple-to-order Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinet Packages, which include: Cabinet; manual, adjustable height stand; factory installed UV light and one set of armrests. These safety cabinets deliver exceptional design and technology advancements such as superior protection with proprietary airflow design, exceptional ergonomics for a safe and comfortable environment, and outstanding energy efficiency for operational cost savings.
Benefits
  • Save money over the entire life of the cabinet with energy efficient design
  • Reduce operating costs up to 75% over traditional biological safety cabinets with AC motors
  • Increase comfort and encourage safe working habits with the sloped front, bright workspace, and low noise level
  • Maximize safety through our innovative Thermo Scientific™ SmartFlow™ design with Digital Airflow Verification
  • Organize the interior with confidence using SmartPort, a clean and easy way to organize with confidence due to the negative pressure dual-side wall
Safety Features:
  • Series meets NSF/ANSI™ Standard 49.
  • Real-time airflow adjustments ensure inflow and downflow velocities remain steady.
  • Pressure sensor filter monitoring measures total airflow across the working area.
  • Proprietary SmartClean front window design simplifies cleaning for a safer working environment.
  • SmartFlow Indicator adjusts airflows as filter resistance changes.
  • SmartFlow visually demonstrates cabinet's ability to compensate to maintain personal protection.
More Choices:
  • Available in 3, 4, 5, or 6' widths (0.9 m, 1.2 m, 1.5 m, or 1.8 m)
  • Available with either 8 or 10 in. work aperture (20.3 or 25.4cm)
  • Available in SmartCoat or stainless-steel interior
  • Easy-to-order package configurations of popular accessories (manual adjustable height stand, UV light, and armrest)
  • Electrical or manual adjustable height stands and Ergolign chair available
Certifications/Compliance:
  • NSF/ANSI 49, UL, CE
Specifications
Description20.3 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)20.3 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEA