accuflex™ Electrode Support
accuflex™ Electrode Support
Actual product may vary
accuflex™ Electrode Support
accuflex™ Electrode Support
accuflex™ Electrode Support
accuflex™ Electrode Support
accuflex™ Electrode Support
accuflex™ Electrode Support
accuflex™ Electrode Support
For accumet™ benchtop meters
Have Questions?
Catalog NumberForUseWithEquipment
13637671
Catalog number 13637671
Price (MYR)
-
For accumet™ benchtop meters
  • Attaches to meter base with bracket supplied
  • Holds one micro electrode and three standard electrodes
  • Permits full up/down and left/right positioning of electrodes
  • Made of chemical-resistant plastic
Specifications
TypeSupport Arm and Bracket
For Use With (Application)pH
IncludesElectrode support arm bracket for meter
MaterialPlastic
Unit SizeEA