Check Order Status
Max Mono II Monocular Biological Microscope
Actual product may vary
Max Mono II Monocular Biological Microscope
Ergonomically designed for everyday use with easily accessible controls for stage movement coarse and fine focus plus varying light intensity.
Have Questions?
Catalog Number
Eyepiece
1201-4000
10X/18 mm
Catalog number
1201-4000
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Eyepiece:
10X/18 mm
Reversed 4-position turret for easy access to stage
With carrying handles
Full mechanical stage
Pre-centered Abbe condenser
With three achromatic objectives – 4x, 10x, 60x
20W illumination and comes with a blue filter and dust cover
Specifications
Condenser Type
Abbe
Eyepiece
10X/18 mm
Head Style
Rotating
Lighting Type
Halogen
Model
Max Mono II
Objective Types
Achromatic
Electrical Requirements
220/240 V 50/60 Hz
Voltage
220/240 V
Frequency
50/60 Hz