T-1250 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
T-1250 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Actual product may vary
T-1250 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Effectively perform a wide range of applications with the Thermo Scientific™ T-1250 Rotor, which offers 12-place capacity.
Have Questions?
Catalog NumberCapacity
1171312 x 36 mL
Catalog number 11713
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
12 x 36 mL
Effectively perform a wide range of applications with the Thermo Scientific™ T-1250 Rotor, which offers 12-place capacity.
Recommended for:
Differential protein fractionation, DNA isolation.
Specifications
DescriptionT-1250 Fixed Angle Rotor
MaterialTitanium
TypeFixed Angle Rotor
Capacity12 x 36 mL
Max. RCF301,032 x g
Max. Speed50,000 rpm
Radius (Metric)108 mm
Radius (Metric) Min.54.7 mm
K Factor68.7
Max. Tube Length97 mm
Max. Tube Diameter25 mm
Net Weight24.5 lb. (11.1 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle24°
Avg. RCF227,030 x g
Min. RCF152,750 x g
Unit SizeEA