n-Decylamine, 99%
n-Decylamine, 99%
n-Decylamine, 99%
Acros Organics
n-Decylamine, 99%
CAS: 2016-57-1 | C10H23N | 157.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 111920100
Price (MYR)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2016-57-1
IUPAC Namedecan-1-amine
Molecular FormulaC10H23N
InChI KeyMHZGKXUYDGKKIU-UHFFFAOYSA-N
SMILESCCCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to yellow to brown
Refractive index1.4350 to 1.4370 (20°C, 589 nm)