Qualitative Ashless Papers; Circle
Product Image
Qualitative Ashless Papers; Circle
Obtain high-quality filtration. Munktell Filtrak™ Qualitative Ashless Papers contain nearly 100% high alpha-cellulose are manufactured under tightly controlled manufacturing conditions to assure lot to lot quality.
Have Questions?
Catalog NumberDiameter (Metric)Particle Retention
110113
also known as R110113
320 mm>10 μm
Catalog number 110113
also known as R110113
Price (MYR)
996.00
EA
Add to cart
Diameter (Metric):
320 mm
Particle Retention:
>10 μm
Price (MYR)
996.00
EA
Add to cart
  • Very fast filtering, open fiber matrix
  • Fast filtering, open to semi-open fiber matrix
  • Medium to fast filtering, semi-open to open fiber matrix
  • Medium to slow filtering, medium-tight fiber matrix
  • Very slow filtration, very tight fiber matrix
Specifications
MaterialCellulose
Particle Retention>10 μm
Filtration Speed11s/10mL
For Use With (Application)For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis
GradeV3
Diameter (Metric)320 mm
Unit SizeEA