Owl™ EC-105 Compact Power Supply
Owl™ EC-105 Compact Power Supply
Actual product may vary
Owl™ EC-105 Compact Power Supply
Carry out DNA and RNA electrophoresis with the help of theThermo Scientific™ Owl™ EC-105 Compact Power Supply
Have Questions?
Catalog NumberVoltage
105ECA-LVD230 V
Catalog number 105ECA-LVD
Price (MYR)
5,110.00
EA
Add to cart
Voltage:
230 V
Price (MYR)
5,110.00
EA
Add to cart
Carry out DNA and RNA electrophoresis with the help of theThermo Scientific™ Owl™ EC-105 Compact Power Supply, equipped with an overcurrent detector for maximum safety.
  • Easy-to-use power supply operates in constant voltage.
  • Adjustment knob and hi-lo range switch control the output, which is variable in 10V increments from 20V to 250V.
  • Two-position switch toggles the bright LED display from indicating output voltage to output current.
  • Supply is equipped with an overcurrent detector for maximum safety and has two jacks.
Specifications
Certifications/ComplianceCE marked
Current300 mA @ 150-250 V; 500 mA below 150 V
DescriptionEC-105 Compact Power Supply; 230V/50Hz
Max. Current500 mA
Product LineOwl
DisplayLED, Voltage or current
No. of Jacks2
Warranty1 Year
Dimensions (L x W x H)6 x 8 x 4.5 in. (15 x 20 x 11 cm)
Electrical Requirements230 V 50 Hz
Max. Voltage230 V
Voltage230 V
Frequency50 Hz
Unit SizeEA