Saf-T Tainer™ Biomedical Waste Container
Saf-T Tainer™ Biomedical Waste Container
Actual product may vary
Saf-T Tainer™ Biomedical Waste Container
A plastic liner sandwiched between corrugated walls makes this Saf-T Tainer™ Biomedical Waste Container leak and puncture resistant.
Have Questions?
Catalog NumberCapacity (English)Diameter (English) OuterHeight (English)
10.20055 gal.12 in.12 in.
Catalog number 10.2005
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (English):
5 gal.
Diameter (English) Outer:
12 in.
Height (English):
12 in.
  • Rectangular, easy-to-assemble container stacks for convenient storage until incinerated
  • Integral handles for safe handling
  • Unassembled containers store flat to conserve space
Specifications
Capacity (Metric)18.9 L
DesignInfectious Waste with Biohazard Symbol
Diameter (Metric) Outer30.48 cm
Diameter (English) Outer12 in.
Capacity (English)5 gal.
ColorRed, Black
Height (Metric)30.48 cm
Width (Metric)30.4 cm
Liner MaterialPlastic
Width (English)30.48 in.
Height (English)12 in.
Product TypeBiomedical Waste Container
Unit SizeEA