Check Order Status
Thermo Scientific Ebmpapst Motor
Thermo Scientific Ebmpapst Motor
Thermo Scientific Ebmpapst motor, PFB, 230V
Have Questions?
Catalog number
097766
Price (MYR)
1,188.00
EA
Add to cart
Price (MYR)
1,188.00
EA
Add to cart
Thermo Scientific Ebmpapst motor, PFB, 230V, 50-60HZ
230V Ebmpapst motor.
Specifications
Type
230 V Motor