Fiberlite F14-14 x 50cy for LYNX 4000 and 6000 Superspeed Centrifuges
Fiberlite F14-14 x 50cy for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
Fiberlite F14-14 x 50cy for LYNX 4000 and 6000 Superspeed Centrifuges
Shorten set-up time and experience peace of mind that the rotor is secure with the Thermo Scientific™ Fiberlite™ F14-14 x 50cy Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
096-14507514 x 50 mL
Catalog number 096-145075
Price (MYR)
37,894.00
EA
Add to cart
Capacity:
14 x 50 mL
Price (MYR)
37,894.00
EA
Add to cart
Shorten set-up time and experience peace of mind that the rotor is secure with the Thermo Scientific™ Fiberlite™ F14-14 x 50cy Fixed-Angle Rotor. Ideal for pelleting cells, tissue homogenates or subcellular separations at high speeds, this carbon fiber rotor supports 50mL conical tubes, with adapters for 15mL or other tubes.
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
Thermo Scientific™ Auto-ID Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Makes lifting and carrying of rotors easier and safer, further enhanced with the lightweight design
Improved Ergonomics and Performance:
  • Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal.
  • Corrosion and fatigue-resistance secures the rotor's structural integrity for unequalled durability.
  • Rotor is backed by a 15-year warranty.
Specifications
DescriptionFiberlite F14-14x50cy Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity14 x 50 mL
Max. RCF33,746 x g
Max. Speed14,000 rpm
Radius (Metric)154 mm
Radius (Metric) Min.83 mm
K Factor798
Max. Tube Length115 mm
Max. Tube Diameter29 mm
Net Weight17.5 lb. (8.0 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle34°
BiocontainmentYes
Unit SizeEA