Fiberlite F20-12 x 50 LEX Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Fiberlite F20-12 x 50 LEX Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
Fiberlite F20-12 x 50 LEX Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Perform subcellular separations using a Thermo Scientific™ Fiberlite™ F20-12 x 50 LEX Fixed-Angle Carbon Fiber Rotor, which features improved ergonomics and advanced performance.
Have Questions?
Catalog NumberCapacity
096-12437512 x 50 mL
Catalog number 096-124375
Price (MYR)
43,359.00
EA
Add to cart
Capacity:
12 x 50 mL
Price (MYR)
43,359.00
EA
Add to cart
Perform subcellular separations using a Thermo Scientific™ Fiberlite™ F20-12 x 50 LEX Fixed-Angle Carbon Fiber Rotor, which features improved ergonomics and advanced performance. The lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal. Corrosion- and fatigue-resistant construction secures the rotor's structural integrity for unequaled durability.
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionFiberlite F20-12x50 LEX Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity12 x 50 mL
Max. RCF51,428 x g
Max. Speed20,000 rpm
Radius (Metric)115 mm
Radius (Metric) Min.56 mm
K Factor455
Max. Tube Length115 mm
Max. Tube Diameter29 mm
Net Weight11.5 lb. (5.25 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle25°
BiocontainmentYes
Unit SizeEA